Product Details

SNP ID
rs199677542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117989520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACCTTACCGCAGTGACCTTGGA[C/T]CTGTACCACAGCAATGGCTACCGGG
Phenotype
MIM: 146933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL10RA PubMed Links

Gene Details

Gene
IL10RA
Gene Name
interleukin 10 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001558.3 344 Silent Mutation GAC,GAT D89D NP_001549.2

View Full Product Details