Product Details

SNP ID
rs199869197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112143732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGTATGATGAAGATTCAAATTGC[A/G]TCTTATTATCATGTCCTGGGACACT
Phenotype
MIM: 600953
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL18 PubMed Links

Gene Details

Gene
IL18
Gene Name
interleukin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243211.1 666 Missense Mutation ACG,ATG T145M NP_001230140.1
NM_001562.3 666 Missense Mutation ACG,ATG T149M NP_001553.1
XM_011542805.1 666 Missense Mutation ACG,ATG T145M XP_011541107.1
XM_011542806.2 666 Missense Mutation ACG,ATG T149M XP_011541108.1

View Full Product Details