Product Details
- SNP ID
-
rs199825108
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:43680962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGAGTGGGGAATGAGGCGGGGGT[C/G]GGCCCGGGGCTCGGAGAATGGGCAG
- Phenotype
-
MIM: 609574
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HSD17B12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56168061] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSD17B12
- Gene Name
- hydroxysteroid 17-beta dehydrogenase 12
View Full Product Details