Product Details

SNP ID
rs199917887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57489360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGCTTTTGGATCTTGCAGTAG[C/T]GTGGTCTGATGGATTTGGTAGCAAC
Phenotype
MIM: 603733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC43A1 PubMed Links

Gene Details

Gene
SLC43A1
Gene Name
solute carrier family 43 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198810.1 1321 Missense Mutation CAC,CGC H409R NP_001185739.1
NM_003627.5 1321 Missense Mutation CAC,CGC H409R NP_003618.1
XM_005274358.4 1321 Missense Mutation CAC,CGC H425R XP_005274415.1
XM_011545320.2 1321 Missense Mutation CAC,CGC H394R XP_011543622.1
XM_011545321.2 1321 Missense Mutation CAC,CGC H153R XP_011543623.1
XM_011545322.1 1321 Missense Mutation CAC,CGC H153R XP_011543624.1
XM_017018451.1 1321 Missense Mutation CAC,CGC H409R XP_016873940.1
XM_017018452.1 1321 Missense Mutation CAC,CGC H378R XP_016873941.1
XM_017018453.1 1321 Missense Mutation CAC,CGC H153R XP_016873942.1

View Full Product Details