Product Details

SNP ID
rs199665374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6211811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTGCCGCCTGCCGGGCCCTGG[C/T]TGGACTGTGTGCATGCCGCAGGAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf42 PubMed Links

Gene Details

Gene
C11orf42
Gene Name
chromosome 11 open reading frame 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173525.2 2951 Intron NP_775796.2
XM_011519926.2 2951 Intron XP_011518228.1
XM_011519927.2 2951 Intron XP_011518229.1
Gene
FAM160A2
Gene Name
family with sequence similarity 160 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098794.1 2951 Missense Mutation ACC,GCC T872A NP_001092264.1
NM_032127.3 2951 Missense Mutation ACC,GCC T886A NP_115503.2
XM_006718343.2 2951 Missense Mutation ACC,GCC T886A XP_006718406.1
XM_011520397.2 2951 Missense Mutation ACC,GCC T886A XP_011518699.1
XM_011520398.2 2951 Intron XP_011518700.1
XM_011520399.2 2951 Intron XP_011518701.1

View Full Product Details