Product Details

SNP ID
rs199874491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14644461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTCTTCAGCATCGCCTGTGCCT[C/T]CTTCTTCCTCACCTGCTTCCTCACC
Phenotype
MIM: 602047 MIM: 602854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE3B PubMed Links

Gene Details

Gene
PDE3B
Gene Name
phosphodiesterase 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000922.3 771 Missense Mutation TCC,TTC S129F NP_000913.2
XM_006718249.3 771 Missense Mutation TCC,TTC S129F XP_006718312.1
XM_017017911.1 771 Missense Mutation TCC,TTC S129F XP_016873400.1
XM_017017912.1 771 Missense Mutation TCC,TTC S129F XP_016873401.1
Gene
PSMA1
Gene Name
proteasome subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143937.1 771 Intron NP_001137409.1
NM_002786.3 771 Intron NP_002777.1
NM_148976.2 771 Intron NP_683877.1

View Full Product Details