Product Details
- SNP ID
-
rs202219612
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:1950914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTGTACAGGTACCCCCTGTACCG[G/T]CTGGGTGGCCCACAACTTCGGGTGT
- Phenotype
-
MIM: 600789
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL23
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34134444] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL23
- Gene Name
- mitochondrial ribosomal protein L23
- Gene
- SNORD131
- Gene Name
- small nucleolar RNA, C/D box 131
There are no transcripts associated with this gene.
View Full Product Details