Product Details

SNP ID
rs202201512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4450101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAGGTGCCATCTTAGCCTTCTA[A/C]ACAACTGTGGTCATCTCTTCAGTCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR52K2 PubMed Links

Gene Details

Gene
OR52K2
Gene Name
olfactory receptor family 52 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005172.2 807 Nonsense Mutation TAA,TAC *254Y NP_001005172.2

View Full Product Details