Product Details
- SNP ID
-
rs202182976
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:64765971 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATACATCATGCCGGCGGAACCAGG[C/T]GGTGGAGGCGGCGGAGGGGGAGGGG
- Phenotype
-
MIM: 608455
MIM: 601516
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PYGM
PubMed Links
Gene Details
- Gene
- PYGM
- Gene Name
- phosphorylase, glycogen, muscle
There are no transcripts associated with this gene.
- Gene
- SF1
- Gene Name
- splicing factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001178030.1 |
1895 |
Intron |
|
|
NP_001171501.1 |
NM_001178031.1 |
1895 |
Silent Mutation |
CCA,CCG |
P563P |
NP_001171502.1 |
NM_004630.3 |
1895 |
Silent Mutation |
CCA,CCG |
P589P |
NP_004621.2 |
NM_201995.2 |
1895 |
Silent Mutation |
CCA,CCG |
P589P |
NP_973724.1 |
NM_201997.2 |
1895 |
Intron |
|
|
NP_973726.2 |
NM_201998.2 |
1895 |
Intron |
|
|
NP_973727.1 |
XM_011545244.1 |
1895 |
Silent Mutation |
CCA,CCG |
P563P |
XP_011543546.1 |
XM_011545245.1 |
1895 |
Missense Mutation |
ACC,GCC |
T565A |
XP_011543547.1 |
XM_011545246.1 |
1895 |
Missense Mutation |
ACC,GCC |
T562A |
XP_011543548.1 |
XM_011545247.1 |
1895 |
Missense Mutation |
ACC,GCC |
T565A |
XP_011543549.1 |
XM_011545248.1 |
1895 |
Intron |
|
|
XP_011543550.1 |
XM_017018244.1 |
1895 |
Silent Mutation |
CCA,CCG |
P563P |
XP_016873733.1 |
XM_017018245.1 |
1895 |
Silent Mutation |
CCA,CCG |
P593P |
XP_016873734.1 |
XM_017018246.1 |
1895 |
Intron |
|
|
XP_016873735.1 |
XM_017018247.1 |
1895 |
Missense Mutation |
ACC,GCC |
T588A |
XP_016873736.1 |
XM_017018248.1 |
1895 |
Intron |
|
|
XP_016873737.1 |
XM_017018249.1 |
1895 |
Silent Mutation |
CCA,CCG |
P474P |
XP_016873738.1 |
XM_017018250.1 |
1895 |
Silent Mutation |
CCA,CCG |
P474P |
XP_016873739.1 |
XM_017018251.1 |
1895 |
Silent Mutation |
CCA,CCG |
P474P |
XP_016873740.1 |
View Full Product Details