Product Details
- SNP ID
-
rs201476503
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:44934962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGACAAAAAGGAGGCGGCTGACCAG[G/T]AGCAGCAGGATCCCCAGGGACATGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TP53I11
PubMed Links
Gene Details
- Gene
- TP53I11
- Gene Name
- tumor protein p53 inducible protein 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001258320.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_001245249.1 |
NM_001258321.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_001245250.1 |
NM_001258322.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_001245251.1 |
NM_001258323.2 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_001245252.1 |
NM_001258324.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_001245253.1 |
NM_001318384.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_001305313.1 |
NM_001318385.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_001305314.1 |
NM_001318386.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_001305315.1 |
NM_001318387.1 |
953 |
Silent Mutation |
CTA,CTC |
L112L |
NP_001305316.1 |
NM_001318388.1 |
953 |
Silent Mutation |
CTA,CTC |
L111L |
NP_001305317.1 |
NM_001318389.1 |
953 |
Silent Mutation |
CTA,CTC |
L89L |
NP_001305318.1 |
NM_001318390.1 |
953 |
Silent Mutation |
CTA,CTC |
L86L |
NP_001305319.1 |
NM_006034.4 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
NP_006025.2 |
XM_005253227.4 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
XP_005253284.1 |
XM_005253229.1 |
953 |
Intron |
|
|
XP_005253286.1 |
XM_011520475.2 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
XP_011518777.1 |
XM_011520476.2 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
XP_011518778.1 |
XM_011520477.2 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
XP_011518779.1 |
XM_011520478.2 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
XP_011518780.1 |
XM_017018580.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
XP_016874069.1 |
XM_017018581.1 |
953 |
Silent Mutation |
CTA,CTC |
L164L |
XP_016874070.1 |
- Gene
- TSPAN18
- Gene Name
- tetraspanin 18
There are no transcripts associated with this gene.
View Full Product Details