Product Details

SNP ID
rs201735019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65540256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCGGGCCTCACCCATCTCTTCC[A/G]GGCTCAGGCACTGGCGCTGCGCGGG
Phenotype
MIM: 602090 MIM: 607982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LTBP3 PubMed Links

Gene Details

Gene
LTBP3
Gene Name
latent transforming growth factor beta binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130144.2 3684 Missense Mutation CCG,CTG P1078L NP_001123616.1
NM_001164266.1 3684 Missense Mutation CCG,CTG P961L NP_001157738.1
NM_021070.4 3684 Missense Mutation CCG,CTG P1078L NP_066548.2
XM_011545032.2 3684 Missense Mutation CCG,CTG P1087L XP_011543334.1
XM_011545033.2 3684 Missense Mutation CCG,CTG P1087L XP_011543335.1
XM_017017737.1 3684 Intron XP_016873226.1
Gene
SCYL1
Gene Name
SCY1 like pseudokinase 1
There are no transcripts associated with this gene.

View Full Product Details