Product Details

SNP ID
rs201530260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112143600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCATGAAATTTTAATAGCTAG[C/T]CTTCGTTTTGAACAGTGAACATTAT
Phenotype
MIM: 600953
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL18 PubMed Links

Gene Details

Gene
IL18
Gene Name
interleukin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243211.1 798 Missense Mutation GAC,GGC D189G NP_001230140.1
NM_001562.3 798 Missense Mutation GAC,GGC D193G NP_001553.1
XM_011542805.1 798 Missense Mutation GAC,GGC D189G XP_011541107.1
XM_011542806.2 798 Missense Mutation GAC,GGC D193G XP_011541108.1

View Full Product Details