Product Details

SNP ID
rs201717789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114405768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGATAATTCAGAGATCTTTCT[C/T]TTCTCCAGAAAATTTTCAGAGACAA
Phenotype
MIM: 612413
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf71 PubMed Links

Gene Details

Gene
C11orf71
Gene Name
chromosome 11 open reading frame 71
There are no transcripts associated with this gene.

Gene
RBM7
Gene Name
RNA binding motif protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286045.1 302 Missense Mutation TCT,TTT S137F NP_001272974.1
NM_001286046.1 302 Missense Mutation TCT,TTT S16F NP_001272975.1
NM_001286047.1 302 Missense Mutation TCT,TTT S16F NP_001272976.1
NM_001286048.1 302 Missense Mutation TCT,TTT S16F NP_001272977.1
NM_016090.3 302 Missense Mutation TCT,TTT S136F NP_057174.1

View Full Product Details