Product Details

SNP ID
rs201467230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124543543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATCAAGCCCAGGTTCCCCACCA[C/T]GGTGACCGTGTAGAAACCCAGAAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8B12 PubMed Links

Gene Details

Gene
OR8B12
Gene Name
olfactory receptor family 8 subfamily B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005195.1 112 Missense Mutation ATG,GTG M38V NP_001005195.1

View Full Product Details