Product Details

SNP ID
rs201491900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60462424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGGACTTTCCCGGCAGAGCCAA[C/T]GAAAGGCCCTATTGCTATGCAATCT
Phenotype
MIM: 112210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MS4A1 PubMed Links

Gene Details

Gene
MS4A1
Gene Name
membrane spanning 4-domains A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021950.3 203 Missense Mutation ACG,ATG T17M NP_068769.2
NM_152866.2 203 Missense Mutation ACG,ATG T17M NP_690605.1

View Full Product Details