Product Details

SNP ID
rs201443300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72758574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATAATGATGTAATCAGCGCCCA[C/T]GGGGAGCCAGGAGCGGAGGGTGATG
Phenotype
MIM: 606646
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARAP1 PubMed Links

Gene Details

Gene
ARAP1
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1
There are no transcripts associated with this gene.

Gene
STARD10
Gene Name
StAR related lipid transfer domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006645.2 1359 Missense Mutation ATG,GTG M139V NP_006636.2

View Full Product Details