Product Details

SNP ID
rs201724564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59457364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAGCCCCTGCACTATGTGACCA[C/T]GATGAGGAAAGAGGTGTGGGTGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4D6 PubMed Links

Gene Details

Gene
OR4D6
Gene Name
olfactory receptor family 4 subfamily D member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004708.1 404 Missense Mutation ACG,ATG T135M NP_001004708.1

View Full Product Details