Product Details

SNP ID
rs201631641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116779774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCAGGGAAGGTCTACTATACCT[G/T]CAAGTAACTGTTTCCTGCTGGATCA
Phenotype
MIM: 606368 MIM: 603901
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APOA5 PubMed Links

Gene Details

Gene
APOA5
Gene Name
apolipoprotein A5
There are no transcripts associated with this gene.

Gene
BUD13
Gene Name
BUD13 homolog
There are no transcripts associated with this gene.

Gene
ZPR1
Gene Name
ZPR1 zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317086.1 1291 Missense Mutation AAG,CAG K361Q NP_001304015.1
NM_003904.4 1291 Missense Mutation AAG,CAG K415Q NP_003895.1

View Full Product Details