Product Details

SNP ID
rs201703612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56276545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATGCTGAATCCCATCATCTAC[A/G]GTTTGCGGAACAAAGATGTAAAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5T1 PubMed Links

Gene Details

Gene
OR5T1
Gene Name
olfactory receptor family 5 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004745.1 907 Missense Mutation AGT,GGT S303G NP_001004745.1

View Full Product Details