Product Details

SNP ID
rs201795636
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4769231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCATTCTTCAGGGGAGGCAAT[C/T]CTGAGGACAGAGTGAATAATTAAGA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR51F1 PubMed Links

Gene Details

Gene
OR51F1
Gene Name
olfactory receptor family 51 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004752.1 687 Silent Mutation AGA,AGG R229R NP_001004752.1

View Full Product Details