Product Details

SNP ID
rs201447269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55967872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATCAACAGAAATGGCACCGTGA[A/T]AAACACCACCGCTACTACATGGACT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10AG1 PubMed Links

Gene Details

Gene
OR10AG1
Gene Name
olfactory receptor family 10 subfamily AG member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005491.1 592 Missense Mutation ATC,TTC I198F NP_001005491.1

View Full Product Details