Product Details

SNP ID
rs201540547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1015814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGATACCGGGGCAGAAGCAGG[G/T]GTGCTTCCATGGGCAGTGAGGGAGC
Phenotype
MIM: 607242 MIM: 158374
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AP2A2 PubMed Links

Gene Details

Gene
AP2A2
Gene Name
adaptor related protein complex 2 alpha 2 subunit
There are no transcripts associated with this gene.

Gene
MUC6
Gene Name
mucin 6, oligomeric mucus/gel-forming
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005961.2 7038 Silent Mutation ACA,ACC T2329T NP_005952.2

View Full Product Details