Product Details

SNP ID
rs201610370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65085200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCAACATACCCCTGGCCAGCC[A/G]AGAGACGACCCGCCTTGTCTGCTAT
Phenotype
MIM: 609374
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDCA5 PubMed Links

Gene Details

Gene
CDCA5
Gene Name
cell division cycle associated 5
There are no transcripts associated with this gene.

Gene
TMEM262
Gene Name
transmembrane protein 262
There are no transcripts associated with this gene.

Gene
ZFPL1
Gene Name
zinc finger protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006782.3 353 Missense Mutation CAA,CGA Q63R NP_006773.2

View Full Product Details