Product Details

SNP ID
rs201497041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4769331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAATTAATCCACAGATGCTATTT[A/G]CCCGAATGTCTGAACATGCTAATTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51F1 PubMed Links

Gene Details

Gene
OR51F1
Gene Name
olfactory receptor family 51 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004752.1 587 Missense Mutation NP_001004752.1

View Full Product Details