Product Details

SNP ID
rs202108761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78016871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCACACAAGCACGCTGGACTTCC[A/G]TGCTGTTATGGCTTCTGCACGTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD14 PubMed Links

Gene Details

Gene
KCTD14
Gene Name
potassium channel tetramerization domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282406.1 1136 Missense Mutation NP_001269335.1
NM_023930.3 1136 Missense Mutation NP_076419.2
Gene
NDUFC2-KCTD14
Gene Name
NDUFC2-KCTD14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203260.1 1136 UTR 3 NP_001190189.1
NM_001203261.1 1136 UTR 3 NP_001190190.1
NM_001203262.1 1136 UTR 3 NP_001190191.1

View Full Product Details