Product Details

SNP ID
rs201927418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64791930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCAGGTGGGATGAGGATGTCGG[A/G]CGTCAGGCCAGCCTCCAGGGGCAGG
Phenotype
MIM: 603166
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP4K2 PubMed Links

Gene Details

Gene
MAP4K2
Gene Name
mitogen-activated protein kinase kinase kinase kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307990.1 2014 Missense Mutation CCC,TCC P683S NP_001294919.1
NM_004579.4 2014 Missense Mutation CCC,TCC P691S NP_004570.2
XM_011545203.2 2014 Missense Mutation CCC,TCC P691S XP_011543505.1
XM_011545204.2 2014 Missense Mutation CCC,TCC P647S XP_011543506.1
XM_017018093.1 2014 Missense Mutation CCC,TCC P639S XP_016873582.1
XM_017018094.1 2014 Missense Mutation CCC,TCC P514S XP_016873583.1
XM_017018095.1 2014 Missense Mutation CCC,TCC P514S XP_016873584.1
XM_017018096.1 2014 Intron XP_016873585.1

View Full Product Details