Product Details

SNP ID
rs202090361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2165722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCACCATAGGCCTTCACCTCCCC[A/G]TTCTGCTTACACAGCCCGAACTCCA
Phenotype
MIM: 176730 MIM: 191290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INS PubMed Links

Gene Details

Gene
INS
Gene Name
insulin
There are no transcripts associated with this gene.

Gene
INS-IGF2
Gene Name
INS-IGF2 readthrough
There are no transcripts associated with this gene.

Gene
MIR4686
Gene Name
microRNA 4686
There are no transcripts associated with this gene.

Gene
TH
Gene Name
tyrosine hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000360.3 3453 Silent Mutation AAC,AAT N382N NP_000351.2
NM_199292.2 3453 Silent Mutation AAC,AAT N413N NP_954986.2
NM_199293.2 3453 Silent Mutation AAC,AAT N409N NP_954987.2
XM_011520335.2 3453 Silent Mutation AAC,AAT N386N XP_011518637.1

View Full Product Details