Product Details

SNP ID
rs201999894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60842753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGACGGTGACAGGCGGCTTCG[C/G]GGCAAAGAAGCGAAAAGGTTCTTCA
Phenotype
MIM: 611293 MIM: 604837
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC86 PubMed Links

Gene Details

Gene
CCDC86
Gene Name
coiled-coil domain containing 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024098.3 Intron NP_077003.1
Gene
PTGDR2
Gene Name
prostaglandin D2 receptor 2
There are no transcripts associated with this gene.

View Full Product Details