Product Details

SNP ID
rs202045268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56318864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTGACAGTCTCCCTTTGTTACC[C/T]TTGCTTTGTTCAAATACACATGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8K3 PubMed Links

Gene Details

Gene
OR8K3
Gene Name
olfactory receptor family 8 subfamily K member 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005202.1 558 Silent Mutation CCC,CCT P186P NP_001005202.1

View Full Product Details