Product Details

SNP ID
rs201814205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46385394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCTCCCGGGCCGCCATCTGCCG[C/T]TTCTTGCGCACCTGGTTGCGAGCGA
Phenotype
MIM: 118495 MIM: 601441 MIM: 162096
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRM4 PubMed Links

Gene Details

Gene
CHRM4
Gene Name
cholinergic receptor muscarinic 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000741.3 1215 Silent Mutation AAA,AAG K388K NP_000732.2
Gene
DGKZ
Gene Name
diacylglycerol kinase zeta
There are no transcripts associated with this gene.

Gene
MDK
Gene Name
midkine (neurite growth-promoting factor 2)
There are no transcripts associated with this gene.

Gene
MIR4688
Gene Name
microRNA 4688
There are no transcripts associated with this gene.

View Full Product Details