Product Details

SNP ID
rs202026391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4594919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATCATCCCAGCCACTTTAAATC[A/C]CATCATCTATGGCATGAGGACCAAA
Phenotype
MIM: 613184
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR52I1 PubMed Links

Gene Details

Gene
OR52I1
Gene Name
olfactory receptor family 52 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005169.1 881 Missense Mutation CAC,CCC H294P NP_001005169.1
Gene
OR52I2
Gene Name
olfactory receptor family 52 subfamily I member 2
There are no transcripts associated with this gene.

Gene
TRIM68
Gene Name
tripartite motif containing 68
There are no transcripts associated with this gene.

View Full Product Details