Product Details

SNP ID
rs201821130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118257268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTGTTAGTCTGTGTCTTCTAA[A/G]TAAACAGAGACCTTTTTCTCTTGGT
Phenotype
MIM: 604873 MIM: 611707
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPZL2 PubMed Links

Gene Details

Gene
MPZL2
Gene Name
myelin protein zero like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005797.3 1013 Silent Mutation TAC,TAT Y210Y NP_005788.1
NM_144765.2 1013 Silent Mutation TAC,TAT Y210Y NP_658911.1
Gene
MPZL3
Gene Name
myelin protein zero like 3
There are no transcripts associated with this gene.

View Full Product Details