Product Details

SNP ID
rs201132519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:109424195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCCTGTTTGGACACAGCTGGC[A/G]AGGGCCTTTTGCAAACGGTGGTACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf87 PubMed Links

Gene Details

Gene
C11orf87
Gene Name
chromosome 11 open reading frame 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207645.3 1069 Missense Mutation AAG,GAG K188E NP_997528.2
XM_011542817.2 1069 Missense Mutation AAG,GAG K335E XP_011541119.1
XM_011542818.2 1069 Missense Mutation AAG,GAG K188E XP_011541120.1

View Full Product Details