Product Details

SNP ID
rs201134554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59835918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGCAGCTCAACCCCCCTCAG[C/G]TGGTTATTTATGGTGTATATGACAG
Phenotype
MIM: 609342
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005142.2 997 Missense Mutation CAC,CAG H321Q NP_005133.2
XM_011544939.2 997 Missense Mutation CAC,CAG H307Q XP_011543241.1

View Full Product Details