Product Details

SNP ID
rs201227416
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57485218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCAACAGGAATCCCAGGAGTG[A/G]GAATAGCAGGAGGCCCAGATTCACC
Phenotype
MIM: 610462 MIM: 603733
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RTN4RL2 PubMed Links

Gene Details

Gene
RTN4RL2
Gene Name
reticulon 4 receptor-like 2
There are no transcripts associated with this gene.

Gene
SLC43A1
Gene Name
solute carrier family 43 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198810.1 1653 Missense Mutation NP_001185739.1
NM_003627.5 1653 Missense Mutation NP_003618.1
XM_005274358.4 1653 Missense Mutation XP_005274415.1
XM_011545320.2 1653 Missense Mutation XP_011543622.1
XM_011545321.2 1653 Missense Mutation XP_011543623.1
XM_011545322.1 1653 Missense Mutation XP_011543624.1
XM_017018451.1 1653 Missense Mutation XP_016873940.1
XM_017018452.1 1653 Missense Mutation XP_016873941.1
XM_017018453.1 1653 Missense Mutation XP_016873942.1

View Full Product Details