Product Details

SNP ID
rs201070254
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112026236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTCCCCCACAATGCAGGCAGG[A/C]ACCATAGCCCGTTGGGAAAAAAAAG
Phenotype
MIM: 610493 MIM: 608770
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DIXDC1 PubMed Links

Gene Details

Gene
DIXDC1
Gene Name
DIX domain containing 1
There are no transcripts associated with this gene.

Gene
DLAT
Gene Name
dihydrolipoamide S-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001931.4 977 Silent Mutation GGA,GGC G106G NP_001922.2
XM_011542647.2 977 Silent Mutation GGA,GGC G106G XP_011540949.1

View Full Product Details