Product Details

SNP ID
rs201163284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3827686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGAGCATTCGAGGTAGCGCAC[A/G]GCGTGGATCTGCTTGGCCAGTGCCT
Phenotype
MIM: 615187 MIM: 179505
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGAP2 PubMed Links

Gene Details

Gene
PGAP2
Gene Name
post-GPI attachment to proteins 2
There are no transcripts associated with this gene.

Gene
RHOG
Gene Name
ras homolog family member G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001665.3 791 Silent Mutation GCC,GCT A151A NP_001656.2
XM_005252916.2 791 Silent Mutation GCC,GCT A151A XP_005252973.1
XM_017017719.1 791 Silent Mutation GCC,GCT A151A XP_016873208.1

View Full Product Details