Product Details

SNP ID
rs201209367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124543495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGAAGTACATGGGAGTGTGCA[C/G]GTGAGAGTTCAGCCCAATCAGGGTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR8B12 PubMed Links

Gene Details

Gene
OR8B12
Gene Name
olfactory receptor family 8 subfamily B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005195.1 160 Missense Mutation CTG,GTG L54V NP_001005195.1

View Full Product Details