Product Details

SNP ID
rs201350477
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5545070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCGAAAGGAGATGCCCATAGCAC[A/T]CTTGATGATGGTCTTGGGAGTCAAG
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR52H1 PubMed Links

Gene Details

Gene
OR52H1
Gene Name
olfactory receptor family 52 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005289.1 454 Missense Mutation AGT,TGT S152C NP_001005289.1

View Full Product Details