Product Details
- SNP ID
-
rs201373734
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:122886086 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGACAGCTTAGACGAGGAGGAAAG[C/G]CCTCGATGGGGAAGCCTGCACGAGA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C11orf63
PubMed Links
Gene Details
- Gene
- C11orf63
- Gene Name
- chromosome 11 open reading frame 63
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024806.3 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
NP_079082.2 |
NM_199124.2 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
NP_954575.1 |
XM_005271680.4 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_005271737.1 |
XM_006718915.2 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_006718978.1 |
XM_011543009.2 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_011541311.1 |
XM_017018338.1 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_016873827.1 |
XM_017018339.1 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_016873828.1 |
XM_017018340.1 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_016873829.1 |
XM_017018341.1 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_016873830.1 |
XM_017018342.1 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_016873831.1 |
XM_017018343.1 |
1121 |
Missense Mutation |
AGC,AGG |
S79R |
XP_016873832.1 |
XM_017018344.1 |
1121 |
Intron |
|
|
XP_016873833.1 |
View Full Product Details