Product Details

SNP ID
rs201237922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32583905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCACCATGAGCGTCCCGGCCTT[C/T]ATCGACATCAGTGAAGAAGATCAGG
Phenotype
MIM: 609641
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3M PubMed Links

Gene Details

Gene
EIF3M
Gene Name
eukaryotic translation initiation factor 3 subunit M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307929.1 139 Silent Mutation TTC,TTT F6F NP_001294858.1
NM_006360.5 139 Silent Mutation TTC,TTT F6F NP_006351.2
XM_011519840.2 139 UTR 5 XP_011518142.1

View Full Product Details