Product Details

SNP ID
rs201172074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67492201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGCAGCCACACCATAGCTGCT[C/T]TTGCCCAAGGCAGCTCTCGGGGGTC
Phenotype
MIM: 605555 MIM: 608794
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIP PubMed Links

Gene Details

Gene
AIP
Gene Name
aryl hydrocarbon receptor interacting protein
There are no transcripts associated with this gene.

Gene
MIR6752
Gene Name
microRNA 6752
There are no transcripts associated with this gene.

Gene
PITPNM1
Gene Name
phosphatidylinositol transfer protein membrane associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130848.1 3809 Silent Mutation NP_001124320.1
NM_004910.2 3809 Silent Mutation NP_004901.2
XM_011545396.2 3809 Missense Mutation XP_011543698.1
XM_017018586.1 3809 Missense Mutation XP_016874075.1

View Full Product Details