Product Details

SNP ID
rs201379359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113690252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCTGGCATGCATCAGCCCTTCC[A/G]TCCAGGTAGCCAGCGCAAAGCATGC
Phenotype
MIM: 606751
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS5 PubMed Links

Gene Details

Gene
TMPRSS5
Gene Name
transmembrane protease, serine 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288749.1 5990 Silent Mutation GAC,GAT D282D NP_001275678.1
NM_001288750.1 5990 Silent Mutation GAC,GAT D351D NP_001275679.1
NM_001288751.1 5990 Silent Mutation GAC,GAT D386D NP_001275680.1
NM_001288752.1 5990 Silent Mutation GAC,GAT D326D NP_001275681.1
NM_030770.3 5990 Silent Mutation GAC,GAT D395D NP_110397.2
XM_017018366.1 5990 Silent Mutation GAC,GAT D165D XP_016873855.1
XM_017018367.1 5990 Silent Mutation GAC,GAT D165D XP_016873856.1

View Full Product Details