Product Details

SNP ID
rs201081591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58204159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATGCATATTTCTGCTGATCTGA[A/G]GAAAGGAACACAAAGTCTTCATACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1S2 PubMed Links

Gene Details

Gene
OR1S2
Gene Name
olfactory receptor family 1 subfamily S member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004459.1 Intron NP_001004459.1

View Full Product Details