Product Details
- SNP ID
-
rs201150293
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:126211505 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGGCTCGGAGCTTCTCCGCTAAT[C/T]TCTGGGCATTTATGGCCGTAGAGGC
- Phenotype
-
MIM: 616587
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM118B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2282580] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM118B
- Gene Name
- family with sequence similarity 118 member B
- Gene
- RPUSD4
- Gene Name
- RNA pseudouridylate synthase domain containing 4
View Full Product Details