Product Details

SNP ID
rs199475670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102840444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTTAAGCTGCTGGGTATTGTCC[G/A]AGACCTCAATCCTTTGGGTGTATGG
Phenotype
MIM: 612349
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PAH PubMed Links

Gene Details

Gene
PAH
Gene Name
phenylalanine hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000277.1 1743 Missense Mutation TCG,TTG S424L NP_000268.1
XM_017019370.1 1743 Intron XP_016874859.1

View Full Product Details