Product Details

SNP ID
rs199564744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69365641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCCTTATATTTTAGGATATGTC[A/C]GCATATGTGAAGAAAATCCAGTTTA
Phenotype
MIM: 602116
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
YEATS4 PubMed Links

Gene Details

Gene
YEATS4
Gene Name
YEATS domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300950.1 443 Intron NP_001287879.1
NM_006530.3 443 Silent Mutation TCA,TCC S60S NP_006521.1

View Full Product Details