Product Details

SNP ID
rs199570843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52839887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGTTCTGCTTGGCCATCCTC[A/G]GAGCAGCCTCCAGCTCGTCCACCTT
Phenotype
MIM: 611159 MIM: 611160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT78 PubMed Links

Gene Details

Gene
KRT78
Gene Name
keratin 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300814.1 827 Missense Mutation CCG,CTG P272L NP_001287743.1
NM_173352.3 827 Missense Mutation CCG,CTG P382L NP_775487.2
XM_011538010.1 827 Missense Mutation CCG,CTG P350L XP_011536312.1
Gene
KRT79
Gene Name
keratin 79
There are no transcripts associated with this gene.

View Full Product Details