Product Details
- SNP ID
-
rs201150918
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52645090 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACCCTTTACAGAGCTAGAACCCC[C/T]ACCTCCAGAGCCATATCCTCCTCCA
- Phenotype
-
MIM: 600194
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT2
PubMed Links
Gene Details
- Gene
- KRT2
- Gene Name
- keratin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000423.2 |
1882 |
Missense Mutation |
AGG,GGG |
R617G |
NP_000414.2 |
View Full Product Details