Product Details
- SNP ID
-
rs201104805
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:130342669 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCGCAGCTGGTGGGCTCCACTGC[C/T]GTGAGTGCTTCACCGTTTCTGACTA
- Phenotype
-
MIM: 605571
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PIWIL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11060834] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PIWIL1
- Gene Name
- piwi like RNA-mediated gene silencing 1
View Full Product Details